Learn More
Bellco Glass, Inc. Big Mouth Clear Vial, 2.0mL Crimp Top - 12x32mm
SDP
Catalog No. 501945358
Encompass_Preferred
Clear vials are manufactured from Type 1 Class A Borosilicate Glass. Amber vials are manufactured from Type1 amber glass. Designed for use with varian and other auto-samplers. MADE IN THE USA.Specifications:Type 1 Class A Borosilicate GlassSize: 12x32mmCap Size: 10-425Case Quantity: 1000
Catalog No. 50-194-5358 Supplier Bellco Glass, Inc. Supplier No. 207000050
Clear vials are manufactured from Type 1 Class A Borosilicate Glass. Amber vials are manufactured from Type1 amber glass. Designed for use with varian and other auto-samplers. MADE IN THE USA.Specifications:Type 1 Class A Borosilicate GlassSize: 12x32mmCap Size: 10-425Case Quantity: 1000