Learn More
MSC ZEP BIG ORANGE-E Degreaser, Liquid, Citrus, 275 gal, 1 Each
Catalog No. 502819337
Encompass
ZEP BIG ORANGE-E Citrus Industrial Degreaser and Graffiti Remover is an all-purpose liquid cleaner supplied in a 275 gal tote, designed for effective industrial degreasing and graffiti removal.
  • Clear, orange liquid with a strong citrus scent
  • Has a pH range of 6.5–7.0
  • Emulsifiable in water
  • Supplied in a 275 gal tote for large-scale applications
Catalog No. 50-281-9337 Supplier MSC Supplier No. 17644048
Add to Cart
Add to Cart
Request a Quote
ZEP BIG ORANGE-E Citrus Industrial Degreaser and Graffiti Remover is an all-purpose liquid cleaner supplied in a 275 gal tote, designed for effective industrial degreasing and graffiti removal.
  • Clear, orange liquid with a strong citrus scent
  • Has a pH range of 6.5–7.0
  • Emulsifiable in water
  • Supplied in a 275 gal tote for large-scale applications