Learn More
MSC ZEP Cleaner and Degreaser, Concentrated, Aerosol, 20 oz, 1 ea
Catalog No. 502870028
Encompass
This cleaner and degreaser comes in a 20 oz aerosol can. It is a concentrated, solvent-based formula with a citrus scent, designed for use in dumpsters and grease traps.
  • Aerosol form
  • Can container type
  • Container size: 20 oz
  • Citrus scent
  • Solvent-based composition
  • Concentrated formula
  • Biodegradable
  • Removes oil
  • Primary chemical: D-Limonene
  • Suitable for dumpsters and grease traps
Catalog No. 50-287-0028 Supplier MSC Supplier No. 11395225
This cleaner and degreaser comes in a 20 oz aerosol can. It is a concentrated, solvent-based formula with a citrus scent, designed for use in dumpsters and grease traps.
  • Aerosol form
  • Can container type
  • Container size: 20 oz
  • Citrus scent
  • Solvent-based composition
  • Concentrated formula
  • Biodegradable
  • Removes oil
  • Primary chemical: D-Limonene
  • Suitable for dumpsters and grease traps