Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC ZEP Vehicle Wash, Heavy Duty, Liquid, 5 gal

Catalog No. 502818221
Encompass

This product is a heavy-duty liquid vehicle wash formulated for thorough cleaning of trucks and trailers, including those with pre-painted aluminum surfaces. It is provided in a 5 gal pail.

  • Designed for heavy-duty truck and trailer washing
  • Liquid form
  • Supplied in a pail container
  • Volume: 5 gal
  • Has a pH value of 13.0–13.5
  • Density: 1.212 g/cm3
  • Soluble in cold and hot water
  • Appearance: clear, dark, brown liquid
  • Characteristic odor

Catalog No. 50-281-8221 Supplier MSC Supplier No. 17731852
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product is a heavy-duty liquid vehicle wash formulated for thorough cleaning of trucks and trailers, including those with pre-painted aluminum surfaces. It is provided in a 5 gal pail.

  • Designed for heavy-duty truck and trailer washing
  • Liquid form
  • Supplied in a pail container
  • Volume: 5 gal
  • Has a pH value of 13.0–13.5
  • Density: 1.212 g/cm3
  • Soluble in cold and hot water
  • Appearance: clear, dark, brown liquid
  • Characteristic odor

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.