Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ BigDye™ Terminator Plus Cycle Sequencing Kit

Catalog No. 4337463
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Reactions
1000 Reactions
24 Reactions
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-374-63 1000 Reactions
43-374-61 24 Reactions
43-374-62 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 43-374-63 Supplier Applied Biosystems™ Supplier No. 4337463
Add to Cart
Add to Cart

The BigDye™ Terminator BDT Plus Cycle Sequencing Kit is a Sanger sequencing master mix designed to enable faster results and improved performance across a broad range of sample types.

The BigDye™ Terminator BDT Plus Cycle Sequencing Kit is a Sanger sequencing master mix designed to enable faster results and improved performance across a broad range of sample types. Built on a newly developed high-performance enzyme, BDT Plus enables more efficient sequencing of challenging templates while maintaining full compatibility with existing BigDye™ workflows and capillary electrophoresis (CE) instruments. Compared to legacy BigDye™ Terminator v3.1 chemistry, BDT Plus helps provide enhanced speed, flexibility, and reliability without requiring workflow changes.

Key Features and Benefits

  • Accelerate sequencing workflows with up to ~50% faster time to results
  • Improve performance on challenging templates using an advanced high-processivity enzyme
  • Maintain existing workflows with full compatibility across current BigDye™ protocols and CE instruments
  • Increase success rates and reduce re-runs for greater laboratory efficiency
  • Flexible Protocol Optimization: Supports implementation of customer-optimized workflows, allowing laboratories to maintain validated protocols and specialized applications

Product Details

The BDT Plus kit incorporates a newly engineered enzyme with increased processivity, allowing for more efficient incorporation of dye-labeled terminators during cycle sequencing. This enhanced enzyme performance improves read quality and consistency, particularly when working with difficult templates such as GC-rich regions, secondary structures, or low-quality samples. The optimized formulation supports robust sequencing across a wider range of sample inputs and conditions.

BDT Plus is designed as a drop-in replacement for existing BigDye™ Terminator chemistries, requiring no changes to established workflows. Researchers can integrate the kit directly into their current cycle sequencing protocols, using the same instruments, plastics, and cleanup methods. This compatibility reduces disruption while enabling immediate performance improvements.

The kit supports a wide range of Sanger sequencing research applications, including assay development, mutation detection, plasmid verification, and confirmatory sequencing. It is well suited for use in clinical research, biopharma, academic, and core laboratory environments, where sample diversity and throughput requirements vary. Its ability to process challenging and heterogeneous sample types makes it particularly valuable for labs seeking consistent results across complex workflows.

By combining faster turnaround times with improved sequencing performance, BDT Plus helps laboratories increase productivity, reduce repeat runs, and maintain confidence in their sequencing data.

TRUSTED_SUSTAINABILITY

Specifications

Format Tube
Isolation Technology Magnetic Bead
Sequencing Type Genome and DNA Sequencing
For Use With (Equipment) SeqStudio™ Genetic Analyzer, SeqStudio™ Flex Series Genetic Analyzer, 3500/3500xL Genetic Analyzer, 3730/3730xl DNA Analyzer Thermal Cyclers: GeneAmp™ PCR System 9700 Dual Series, Veriti™ Fast 96‐well and 384‐well Thermal Cyclers
For Use With (Application) Sanger Sequencing, AT-Rich Sequencing (>65%), Comparative Sequencing (Germline Mutations - 50:50), GC-Rich Sequencing (>65%), Mixed-Base Detection, Long-Read Sequencing, Resequencing, De Novo Sequencing - High Throughput (cDNA), GT-Rich or Difficult Template Sequencing
Product Line BigDye Terminator
Product Type Sequencing Kit
Quantity 1000 Reactions
Technique Fluorescent Dye Terminator Sequencing
Template Compatibility BAC DNA, Lambda DNA, Genomic DNA (Bacterial), Single Stranded DNA, Rolling Circle Amplified Products, Fosmids, PCR Amplicons (50:50 Heterozygotes), Plasmid DNA (≤15 Kb), PCR Amplicons

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.