Learn More
Medchemexpress LLC Bigelovin 10mg | 3668-14-2 | 304.34 | C17H20O5 | 10 MG
SDP
Catalog No. 5000226459
Encompass_Preferred
Bigelovin is a sesquiterpene lactone (CAS 3668-14-2; C17H20O5; MW 304.34) that acts as a selective retinoid X receptor α (RXRα) agonist. It suppresses tumor growth by inducing apoptosis and autophagy via inhibition of the mTOR pathway and modulation of reactive oxygen species (ROS). Supplied for research use with documented solubility and storage guidance.
  • Selective retinoid X receptor α (RXRα) agonist activity.
  • Induces apoptosis and autophagy in cancer model studies.
  • Available in small research quantities suitable for in vitro and in vivo formulation.
  • Includes detailed solubility and stock preparation guidance.
  • Storage guidance provided: protect from light at 4°C; solvent storage conditions included.
Catalog No. 50-002-26459 Supplier Medchemexpress LLC Supplier No. HY11650610MG
Bigelovin is a sesquiterpene lactone (CAS 3668-14-2; C17H20O5; MW 304.34) that acts as a selective retinoid X receptor α (RXRα) agonist. It suppresses tumor growth by inducing apoptosis and autophagy via inhibition of the mTOR pathway and modulation of reactive oxygen species (ROS). Supplied for research use with documented solubility and storage guidance.
  • Selective retinoid X receptor α (RXRα) agonist activity.
  • Induces apoptosis and autophagy in cancer model studies.
  • Available in small research quantities suitable for in vitro and in vivo formulation.
  • Includes detailed solubility and stock preparation guidance.
  • Storage guidance provided: protect from light at 4°C; solvent storage conditions included.