Learn More
Medchemexpress LLC Bigelovin 5mg | 3668-14-2 | 304.34 g/mol | C17H20O5 | 5 MG
SDP
Catalog No. 5000226772
Encompass_Preferred
Bigelovin is a sesquiterpene lactone research compound used to induce apoptosis and autophagy through reactive oxygen species-mediated inhibition of mTOR signaling. It is supplied for in vitro research use and is provided in small, high-purity quantities with specified storage conditions to preserve stability.
  • Induces apoptosis and autophagy via ROS-mediated mTOR pathway inhibition.
  • High purity suitable for in vitro research (product page lists 99.92%).
  • Molecular formula C17H20O5; molecular weight 304.34 g/mol.
  • Provided in small-quantity vials appropriate for cell-based assays and biochemical studies.
  • Store protected from light; refrigerated storage recommended; in solution store at low temperatures per datasheet.
Catalog No. 50-002-26772 Supplier Medchemexpress LLC Supplier No. HY1165065MG
Bigelovin is a sesquiterpene lactone research compound used to induce apoptosis and autophagy through reactive oxygen species-mediated inhibition of mTOR signaling. It is supplied for in vitro research use and is provided in small, high-purity quantities with specified storage conditions to preserve stability.
  • Induces apoptosis and autophagy via ROS-mediated mTOR pathway inhibition.
  • High purity suitable for in vitro research (product page lists 99.92%).
  • Molecular formula C17H20O5; molecular weight 304.34 g/mol.
  • Provided in small-quantity vials appropriate for cell-based assays and biochemical studies.
  • Store protected from light; refrigerated storage recommended; in solution store at low temperatures per datasheet.