Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse BIGH3 (TGFBI) ELISA Kit

Catalog No. EMTGFBI
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EMTGFBI 96 Tests
EMTGFBIX5 5 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EMTGFBI Supplier Invitrogen™ Supplier No. EMTGFBI
Add to Cart
Add to Cart

ELISA

The Mouse BIGH3 (TGFBI) ELISA quantitates Ms BIGH3 in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms BIGH3. Principle of the method The Mouse BIGH3 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

This gene encodes an RGD-containing protein that binds to type I, II and IV collagens. The RGD motif is found in many extracellular matrix proteins modulating cell adhesion and serves as a ligand recognition sequence for several integrins. This protein plays a role in cell-collagen interactions and may be involved in endochondrial bone formation in cartilage. The protein is induced by transforming growth factor-beta and acts to inhibit cell adhesion. Mutations in this gene are associated with multiple types of corneal dystrophy.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P82198
Assay Range 3-1000 ng/mL
Assay Sensitivity 3 pg/mL
Calibration Range 3 pg/mL to 1000 ng/mL
Content And Storage Pre-coated plate(s), standard or calibrator, detector antibody, HRP conjugate, diluents, wash buffer, substrate(s), and stop solution. Store kit at 2-8°C. See product manual for detailed contents and storage conditions for maximum stability.
Assay Solid-phase sandwich ELISA
Conjugate HRP
Detection Method Colorimetric
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family Other Proteins
Sample Type Plasma, Serum, Supernatant
Specificity This ELISA pair antibody detects mouse beta-IGH3. Other species not determined.
Sufficient For 96 assays
Target Species Mouse
For Use With (Application) ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias 68kDa; AI181842; AI747162; beta ig-h3; Beta-ig; beta-ig-h3; betaig-h3; BIGH3; Big-h3; CDB1; CDG2; CDGG1; CSD; CSD1; CSD2; CSD3; EBMD; kerato-epithelin; LCD1; MP70; MP78; RGD-CAP; RGD-containing collagen-associated protein; TGFBI; transforming growth factor beta induced; transforming growth factor beta-induced 68kDa; transforming growth factor induced protein; transforming growth factor, beta induced; transforming growth factor, beta induced, 68 kDa; transforming growth factor, beta-induced, 68kD; transforming growth factor, beta-induced, 68kDa; Transforming growth factor-beta-induced protein ig-h368kDa; AI181842; AI747162; beta ig-h3; Beta-ig; beta-ig-h3; betaig-h3; BIGH3; Big-h3; CDB1; CDG2; CDGG1; CSD; CSD1; CSD2; CSD3; EBMD; kerato-epithelin; LCD1; MP70; MP78; RGD-CAP; RGD-containing collagen-associated protein; TGFBI; transforming growth factor beta induced; transforming growth factor beta-induced 68kDa; transforming growth factor induced protein; transforming growth factor, beta induced; transforming growth factor, beta induced, 68 kDa; transforming growth factor, beta-induced, 68kD; transforming growth factor, beta-induced, 68kDa; Transforming growth factor-beta-induced protein ig-h3; 68kDa; AI181842; AI747162; Beta-ig; big-h3
Gene ID (Entrez) 21810
Gene Symbol TGFBI
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Protein Aliases Beta ig-h3
Quantity 96 Tests
Regulatory Status RUO
Research Discipline Angiogenesis
Sample Volume Plasma 5 μL, Serum 5 μL, Supernatant 100 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Synonym 68 kDa, AI181842, AI747162, Beta-ig, big-h3
Target TGFBI
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.