Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Biguanide | 56-03-1 | 98.1% | 10 MG
SDP

Catalog No. 5000234100
Encompass_Preferred

Biguanide (CAS 56-03-1) is a small-molecule biguanide base used as a research reagent. It is reported to inhibit mitochondrial ATP production, activate the LKB1-AMPK signaling pathway, and modulate insulin-receptor signaling; it is commonly used in metabolic and diabetes-related studies.

  • Small-molecule biguanide reagent for research use
  • CAS number 56-03-1
  • Molecular formula C2H7N5; molecular weight 101.11 g/mol
  • Reported to inhibit mitochondrial ATP production and activate AMPK signaling
  • Used in metabolic, insulin signaling, and mitochondrial studies
  • Available in a 10 MG vial; verify pack sizes with supplier

Catalog No. 50-002-34100 Supplier Medchemexpress LLC Supplier No. HY12100610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Biguanide (CAS 56-03-1) is a small-molecule biguanide base used as a research reagent. It is reported to inhibit mitochondrial ATP production, activate the LKB1-AMPK signaling pathway, and modulate insulin-receptor signaling; it is commonly used in metabolic and diabetes-related studies.

  • Small-molecule biguanide reagent for research use
  • CAS number 56-03-1
  • Molecular formula C2H7N5; molecular weight 101.11 g/mol
  • Reported to inhibit mitochondrial ATP production and activate AMPK signaling
  • Used in metabolic, insulin signaling, and mitochondrial studies
  • Available in a 10 MG vial; verify pack sizes with supplier

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.