Learn More
Medchemexpress LLC Biguanide | 56-03-1 | 98.1% | 100 MG
SDP
Catalog No. 5000232729
Encompass_Preferred
Biguanide is an orally active antihyperglycemic research reagent used to investigate mitochondrial metabolism and insulin signaling. It inhibits mitochondrial ATP production and activates the LKB1-AMPK pathway, altering cellular energy homeostasis.
  • Used to study mitochondrial metabolism and AMPK signaling.
  • Inhibits mitochondrial ATP production.
  • Enhances insulin-receptor activation and downstream signaling.
  • Typical purity 98.1% for research use.
  • Provided as a solid suitable for in vitro and in vivo model studies.
Catalog No. 50-002-32729 Supplier Medchemexpress LLC Supplier No. HY121006100MG
Biguanide is an orally active antihyperglycemic research reagent used to investigate mitochondrial metabolism and insulin signaling. It inhibits mitochondrial ATP production and activates the LKB1-AMPK pathway, altering cellular energy homeostasis.
  • Used to study mitochondrial metabolism and AMPK signaling.
  • Inhibits mitochondrial ATP production.
  • Enhances insulin-receptor activation and downstream signaling.
  • Typical purity 98.1% for research use.
  • Provided as a solid suitable for in vitro and in vivo model studies.