Learn More
Medchemexpress LLC Biguanide | 56-03-1 | 98.1% | 25 MG
SDP
Catalog No. 5000234050
Encompass_Preferred
Biguanide is a small-molecule antihyperglycemic research reagent (CAS 56-03-1) that inhibits mitochondrial ATP production, activates the LKB1-AMPK signaling pathway, and enhances insulin-receptor signaling. It is intended for research use only.
  • Antihyperglycemic activity; modulates mitochondrial metabolism.
  • Activates LKB1-AMPK signaling and enhances insulin receptor signaling.
  • Molecular formula C2H7N5; molecular weight 101.11 g/mol.
  • Supplied as a solid in small pack sizes for research applications.
  • For research use only; not for human therapeutic use.
Catalog No. 50-002-34050 Supplier Medchemexpress LLC Supplier No. HY12100625MG
Biguanide is a small-molecule antihyperglycemic research reagent (CAS 56-03-1) that inhibits mitochondrial ATP production, activates the LKB1-AMPK signaling pathway, and enhances insulin-receptor signaling. It is intended for research use only.
  • Antihyperglycemic activity; modulates mitochondrial metabolism.
  • Activates LKB1-AMPK signaling and enhances insulin receptor signaling.
  • Molecular formula C2H7N5; molecular weight 101.11 g/mol.
  • Supplied as a solid in small pack sizes for research applications.
  • For research use only; not for human therapeutic use.