Learn More
Medchemexpress LLC Biguanide | 56-03-1 | 98.1% | 5 MG
SDP
Catalog No. 5000234052
Encompass_Preferred
Biguanide (CAS 56-03-1) is a small-molecule antihyperglycemic agent for research use. It inhibits mitochondrial ATP production, activates the LKB1-AMPK signaling pathway, disrupts cellular energy homeostasis, and enhances insulin receptor signaling. Supplied as a powder with recommended storage conditions for powder and solvent forms.
  • Small-molecule antihyperglycemic agent for research use.
  • Inhibits mitochondrial ATP production and activates the LKB1-AMPK pathway.
  • Used to study insulin receptor signaling and cellular energy homeostasis.
  • Supplied as a powder; powder storage -20°C (stable ~3 years); in solvent store -80°C (6 months) or -20°C (1 month).
  • Molecular formula C2H7N5; molecular weight 101.11 g/mol.
Catalog No. 50-002-34052 Supplier Medchemexpress LLC Supplier No. HY1210065MG
Biguanide (CAS 56-03-1) is a small-molecule antihyperglycemic agent for research use. It inhibits mitochondrial ATP production, activates the LKB1-AMPK signaling pathway, disrupts cellular energy homeostasis, and enhances insulin receptor signaling. Supplied as a powder with recommended storage conditions for powder and solvent forms.
  • Small-molecule antihyperglycemic agent for research use.
  • Inhibits mitochondrial ATP production and activates the LKB1-AMPK pathway.
  • Used to study insulin receptor signaling and cellular energy homeostasis.
  • Supplied as a powder; powder storage -20°C (stable ~3 years); in solvent store -80°C (6 months) or -20°C (1 month).
  • Molecular formula C2H7N5; molecular weight 101.11 g/mol.