Learn More
Grainger BriskHeat Heating Tape, Fiberglass, 0.50 in x 2 ft, 105 W, 1 ea
Catalog No. 502769754
Encompass_Preferred
Heavy Insulated Heating Tapes are designed for laboratory heating applications including apparatus, gas tubing, and valves. Featuring a multi-stranded resistance wire heating element within a fiberglass outer sheath, these tapes provide rapid thermal response and accurate heat delivery. They are compatible with glass and electrically conductive metal surfaces. A temperature controller is required for safe operation.
  • Features a built-in split-plug design
  • Constructed from fiberglass for durability and insulation
  • Offers exceptional flexibility for versatile installation
  • Includes 230 mm (9 in.) long high temperature tie downs
  • Rapid thermal response
  • Maximum exposure temperature of up to 500°C (932°F) for single installation
  • Power density of 1.3 watts/cm² (8.6 watts/in.²)
Catalog No. 50-276-9754 Supplier Grainger Supplier No. 13P748
Heavy Insulated Heating Tapes are designed for laboratory heating applications including apparatus, gas tubing, and valves. Featuring a multi-stranded resistance wire heating element within a fiberglass outer sheath, these tapes provide rapid thermal response and accurate heat delivery. They are compatible with glass and electrically conductive metal surfaces. A temperature controller is required for safe operation.
  • Features a built-in split-plug design
  • Constructed from fiberglass for durability and insulation
  • Offers exceptional flexibility for versatile installation
  • Includes 230 mm (9 in.) long high temperature tie downs
  • Rapid thermal response
  • Maximum exposure temperature of up to 500°C (932°F) for single installation
  • Power density of 1.3 watts/cm² (8.6 watts/in.²)