Learn More
Abcam BIIB 021, 25MG
Catalog No. 502682924
Encompass_Preferred
MW 318.76 Da, Purity >98%. Selective, Hsp90 competitive inhibitor (Ki = 1.7 nM). Displays no significant activity at a range of ATP-binding kinases or Na+K+ ATPase. Induces degradation of HER-2 in vitro (EC₅₀ = 38 nM in MCF-7 cells). Inhibits growth and promotes cell death in a variety of human tumor cells. Orally bioavailable.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-268-2924 Supplier Abcam Supplier No. AB27359925MG
MW 318.76 Da, Purity >98%. Selective, Hsp90 competitive inhibitor (Ki = 1.7 nM). Displays no significant activity at a range of ATP-binding kinases or Na+K+ ATPase. Induces degradation of HER-2 in vitro (EC₅₀ = 38 nM in MCF-7 cells). Inhibits growth and promotes cell death in a variety of human tumor cells. Orally bioavailable.
The product is subject to the following: Abcam Restricted Use Statement