Learn More
Selleck Chemical LLC BIIB021 50mg 848695-25-0 CNF2024
SDP
Catalog No. 508451
Encompass_Preferred
BIIB021 (CNF2024) is an orally available, fully synthetic small-molecule inhibitor of HSP90 with Ki and EC50 of 1.7 nM and 38 nM, respectively. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-845-1 Supplier Selleck Chemical LLC Supplier No. S117550MG
BIIB021 (CNF2024) is an orally available, fully synthetic small-molecule inhibitor of HSP90 with Ki and EC50 of 1.7 nM and 38 nM, respectively. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.