Learn More
Medchemexpress LLC HY-131342 5mg Medchemexpress, BIIB068 CAS:1798787-27-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1313425MG
Encompass_Preferred
Medchemexpress, HY-131342 5mg BIIB068 CAS:1798787-27-5 BIIB068 is a potent, selective, reversible and orally active BTK inhibitor with an IC50 of 1 nM and a Kd of 0.3 nM. BIIB068 shows more >400-fold selective for BTK than other kinases. BIIB068 has the potential for autoimmune diseases research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1276