Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BIIE-0246 | 246146-55-4 | 99.1% | 896.05 g·mol⁻¹ | C49H57N11O6 | 1 ML
SDP

Catalog No. 5000247240
Encompass_Preferred

BIIE-0246 is a potent, selective non-peptide antagonist of the neuropeptide Y Y2 receptor (NPY2R) used in preclinical neuroscience research. It has an IC50 of approximately 15 nM (rat [125I]PYY3-36) and modulates downstream signaling pathways including p-AKT and p44/42 MAPK. The compound is supplied as a 10 mM solution in DMSO with reported high purity for research use only.

  • Potent NPY Y2 receptor antagonist (IC50 ~15 nM).
  • Ready-to-use 10 mM solution in DMSO for in vitro assays.
  • High reported purity (99.1%).
  • Molecular weight 896.05 g·mol⁻¹ and formula C49H57N11O6.
  • Suitable for studies of NPY2R signaling and related pathways.

Catalog No. 50-002-47240 Supplier Medchemexpress LLC Supplier No. HY10198610MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

BIIE-0246 is a potent, selective non-peptide antagonist of the neuropeptide Y Y2 receptor (NPY2R) used in preclinical neuroscience research. It has an IC50 of approximately 15 nM (rat [125I]PYY3-36) and modulates downstream signaling pathways including p-AKT and p44/42 MAPK. The compound is supplied as a 10 mM solution in DMSO with reported high purity for research use only.

  • Potent NPY Y2 receptor antagonist (IC50 ~15 nM).
  • Ready-to-use 10 mM solution in DMSO for in vitro assays.
  • High reported purity (99.1%).
  • Molecular weight 896.05 g·mol⁻¹ and formula C49H57N11O6.
  • Suitable for studies of NPY2R signaling and related pathways.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.