Learn More
Medchemexpress LLC BIIE-0246 | 246146-55-4 | 99.1% | 896.05 g/mol | C49H57N11O6 | 10 MG
SDP
Catalog No. 5000248013
Encompass_Preferred
BIIE-0246 is a non-peptide neuropeptide Y (NPY) Y2 receptor antagonist supplied for laboratory research use. The compound is provided as a high-purity research chemical (CAS 246146-55-4) for in vitro and in vivo pharmacological studies of NPY signaling and receptor pharmacology.
  • Potent and selective NPY Y2 receptor antagonist suitable for receptor pharmacology studies.
  • High purity (99.10%) appropriate for biochemical and cellular assays.
  • Molecular weight 896.05 g/mol, convenient for mass-based preparations.
  • Available in small research pack sizes to support pilot and exploratory studies.
  • Intended for research use only; not for human or clinical applications.
Catalog No. 50-002-48013 Supplier Medchemexpress LLC Supplier No. HY10198610MG
BIIE-0246 is a non-peptide neuropeptide Y (NPY) Y2 receptor antagonist supplied for laboratory research use. The compound is provided as a high-purity research chemical (CAS 246146-55-4) for in vitro and in vivo pharmacological studies of NPY signaling and receptor pharmacology.
  • Potent and selective NPY Y2 receptor antagonist suitable for receptor pharmacology studies.
  • High purity (99.10%) appropriate for biochemical and cellular assays.
  • Molecular weight 896.05 g/mol, convenient for mass-based preparations.
  • Available in small research pack sizes to support pilot and exploratory studies.
  • Intended for research use only; not for human or clinical applications.