Learn More
Medchemexpress LLC Biie-0246 (AR-H 053591) | 246146-55-4 | 99.1% | 896.05 | C49H57N11O6 | 25 MG
SDP
Catalog No. 5000248012
Encompass_Preferred
BIIE-0246 is a selective, non-peptide antagonist of the neuropeptide Y Y2 receptor used in pharmacological research. It exhibits potent activity (IC50 ≈ 15 nM) and modulates NPY-mediated signaling pathways, including effects on p-AKT S473 and p44/42 MAPK; it has been used in disease models such as ADR nephropathy.
  • High potency at NPY Y2 receptor (IC50 ≈ 15 nM).
  • High purity suitable for research applications (99.1%).
  • Available as solid and as DMSO solution for flexible dosing.
  • Molecular formula C49H57N11O6 and molecular weight 896.05.
  • Applicable for in vitro and in vivo studies of NPY signaling and renal pathology.
Catalog No. 50-002-48012 Supplier Medchemexpress LLC Supplier No. HY10198625MG
BIIE-0246 is a selective, non-peptide antagonist of the neuropeptide Y Y2 receptor used in pharmacological research. It exhibits potent activity (IC50 ≈ 15 nM) and modulates NPY-mediated signaling pathways, including effects on p-AKT S473 and p44/42 MAPK; it has been used in disease models such as ADR nephropathy.
  • High potency at NPY Y2 receptor (IC50 ≈ 15 nM).
  • High purity suitable for research applications (99.1%).
  • Available as solid and as DMSO solution for flexible dosing.
  • Molecular formula C49H57N11O6 and molecular weight 896.05.
  • Applicable for in vitro and in vivo studies of NPY signaling and renal pathology.