Learn More
Medchemexpress LLC BIIE-0246 (AR-H 053591) | 246146-55-4 | 99.1% | 896.05 g/mol | C49H57N11O6 | 100 MG
SDP
Catalog No. 5000245906
Encompass_Preferred
BIIE-0246 (AR-H 053591) is a potent, selective non-peptide antagonist of the neuropeptide Y Y2 receptor used as a research reagent to probe NPY2R-mediated signaling. It is provided as a solid powder for in vitro and in vivo applications, with a reported IC50 of approximately 15 nM (rat [125I]PYY3-36). Intended for research use only.
  • Potent, selective NPY Y2 receptor antagonist (IC50 ≈ 15 nM).
  • Supplied as a solid powder suitable for in vitro and in vivo studies.
  • High purity (reported ≥99.1%).
  • Available in multiple pack sizes and solution formats for convenience.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-45906 Supplier Medchemexpress LLC Supplier No. HY101986100MG
BIIE-0246 (AR-H 053591) is a potent, selective non-peptide antagonist of the neuropeptide Y Y2 receptor used as a research reagent to probe NPY2R-mediated signaling. It is provided as a solid powder for in vitro and in vivo applications, with a reported IC50 of approximately 15 nM (rat [125I]PYY3-36). Intended for research use only.
  • Potent, selective NPY Y2 receptor antagonist (IC50 ≈ 15 nM).
  • Supplied as a solid powder suitable for in vitro and in vivo studies.
  • High purity (reported ≥99.1%).
  • Available in multiple pack sizes and solution formats for convenience.
  • Intended for research use only; not for human or clinical use.