Learn More
Medchemexpress LLC BIIE-0246 hydrochloride | 246146-55-4 | MFCD04113026 | >98.0% | 896.0 g/mol | C49H57N11O6 | 5 MG
SDP
Catalog No. 5000247981
Encompass_Preferred
BIIE-0246 hydrochloride is a selective, non-peptide antagonist of the neuropeptide Y Y2 receptor supplied as the hydrochloride salt for research use. It is used in pharmacological studies to probe NPY-mediated signaling and is provided at high purity for in vitro and in vivo experiments.
  • Selective NPY Y2 receptor antagonist
  • High purity (>98.0%) suitable for research
  • Hydrochloride salt for improved aqueous solubility
  • Molecular formula C49H57N11O6; molar mass 896.0 g/mol
  • Available in small research pack sizes, for example 5 MG
Catalog No. 50-002-47981 Supplier Medchemexpress LLC Supplier No. HY1019865MG
BIIE-0246 hydrochloride is a selective, non-peptide antagonist of the neuropeptide Y Y2 receptor supplied as the hydrochloride salt for research use. It is used in pharmacological studies to probe NPY-mediated signaling and is provided at high purity for in vitro and in vivo experiments.
  • Selective NPY Y2 receptor antagonist
  • High purity (>98.0%) suitable for research
  • Hydrochloride salt for improved aqueous solubility
  • Molecular formula C49H57N11O6; molar mass 896.0 g/mol
  • Available in small research pack sizes, for example 5 MG