Learn More
Enzo Life Sciences betaIII-Tubulin (human) monoclonal antibody (TU-20) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804405C100
Encompass_Preferred
Clone: TU-20. Isotype: IgG1. Immunogen: Synthetic peptide corresponding to aa 441-448 (E441SESQGPK448) of human βIII-tubulin (class III β-tubulin). Species reactivity: Human. Specificity: Recognizes βIII-tubulin located in neurons. Applications: ELISA, Flow Cytometry, ICC, IHC (FS), IHC (PS), WB. Formulation: Liquid. In PBS, pH 7.4, containing 15 mM sodium azide.
Catalog No. 50-200-9904