Learn More
Medchemexpress LLC HY-114641A 1mg Medchemexpress, BIIL-260 (hydrochloride) CAS:192581-24-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY114641A1MG
Encompass_Preferred
Medchemexpress, HY-114641A 1mg BIIL-260 (HCl) CAS:192581-24-1 BIIL-260 hydrochloride is a potent and long-acting orally active leukotriene B(4) receptor LTB4 antagonist, with anti-inflammatory activity. BIIL-260 hydrochloride interacts with the LTB4 receptor in a saturable, reversible, and competitive manner, has high affinity to the LTB4 receptor on isolated human neutrophil cell membranes with Ki values of 1.7 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9367