Learn More
Medchemexpress LLC BIIL-260 hydrochlori | 100MG
SDP
Catalog No. 5000228498
Encompass_Preferred
BIIL-260 hydrochloride is a potent and long-acting orally active leukotriene B(4) receptor LTB4 antagonist with anti-inflammatory activity BIIL-260 hydrochloride interacts with the LTB4 receptor in a saturable reversible and competitive manner has high affinity to the LTB4 receptor on isolated human neutrophil cell membranes with Ki values of 1 7 nM[1]
Catalog No. 50-002-28498 Supplier Medchemexpress LLC Supplier No. HY114641A100MG
BIIL-260 hydrochloride is a potent and long-acting orally active leukotriene B(4) receptor LTB4 antagonist with anti-inflammatory activity BIIL-260 hydrochloride interacts with the LTB4 receptor in a saturable reversible and competitive manner has high affinity to the LTB4 receptor on isolated human neutrophil cell membranes with Ki values of 1 7 nM[1]