Learn More
Medchemexpress LLC BIIL-260 hydrochlori 10mg | 192581-24-1 | 10 MG
SDP
Catalog No. 5000229136
Encompass_Preferred
Biil-260 hydrochloride is the hydrochloride salt of BIIL-260, a potent, orally active leukotriene B4 (LTB4) receptor antagonist with anti-inflammatory activity. It binds the human LTB4 receptor reversibly and competitively (reported Ki ≈ 1.7 nM) and is supplied as a high-purity solid for research use.
  • Potent LTB4 receptor antagonist with reported high affinity (Ki ≈ 1.7 nM).
  • High purity solid suitable for in vitro and preclinical research.
  • Available in small milligram quantities for compound screening and assay development.
  • Soluble in DMSO for preparation of concentrated stock solutions.
Catalog No. 50-002-29136 Supplier Medchemexpress LLC Supplier No. HY114641A10MG
Biil-260 hydrochloride is the hydrochloride salt of BIIL-260, a potent, orally active leukotriene B4 (LTB4) receptor antagonist with anti-inflammatory activity. It binds the human LTB4 receptor reversibly and competitively (reported Ki ≈ 1.7 nM) and is supplied as a high-purity solid for research use.
  • Potent LTB4 receptor antagonist with reported high affinity (Ki ≈ 1.7 nM).
  • High purity solid suitable for in vitro and preclinical research.
  • Available in small milligram quantities for compound screening and assay development.
  • Soluble in DMSO for preparation of concentrated stock solutions.