Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BIIL-260 hydrochlori 10mM 1mL | 192581-24-1 | 1 ML
SDP

Catalog No. 5000228499
Encompass_Preferred

Biil-260 hydrochloride is a potent leukotriene B4 (LTB4) receptor antagonist used in research to investigate inflammatory signaling pathways. It is typically supplied at high purity and commonly provided as a 10 mM solution in DMSO for in vitro applications.

  • Potent LTB4 receptor antagonist
  • High purity (99.4%)
  • Molecular weight 503.03 g/mol
  • Supplied as a 10 mM solution in DMSO, 1 mL
  • Appearance: solid, white to off-white
  • Storage: solids sealed at 4°C; in solution store at -80°C for long term

Catalog No. 50-002-28499 Supplier Medchemexpress LLC Supplier No. HY114641A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Biil-260 hydrochloride is a potent leukotriene B4 (LTB4) receptor antagonist used in research to investigate inflammatory signaling pathways. It is typically supplied at high purity and commonly provided as a 10 mM solution in DMSO for in vitro applications.

  • Potent LTB4 receptor antagonist
  • High purity (99.4%)
  • Molecular weight 503.03 g/mol
  • Supplied as a 10 mM solution in DMSO, 1 mL
  • Appearance: solid, white to off-white
  • Storage: solids sealed at 4°C; in solution store at -80°C for long term

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.