Learn More
AdipoGen Bikaverin (250 µg)
SDP
Supplier:  AdipoGen AGCN20130C250
Encompass_Preferred
Chemical. CAS: 33390-21-5. Formula: C20H14O8. MW: 382.3. Isolated from Fusarium sp. Antibiotic. Antiprotozoal. Antifungal. Anticancer. ATP synthesis inhibitior. Haemolytic agent. Antioomycete. Spermidine-induced autoactivation inhibitor. Plasma hyaluronan-binding protein (PHBP) inhibitor (active form). Review.
Catalog No. 50-205-3503