Learn More
Abcam Bikaverin, ATP synthesis inhibitor, 250UG
Catalog No. 502705994
Encompass_Preferred
MW 382.3 Da, Purity >95%. ATP synthesis inhibitor. Antiprotozoal, antibiotic and antifungal agent. Shows antitumor effects against different cancer cell lines and neuroprotective effects.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5994 Supplier Abcam Supplier No. AB144205250UG
MW 382.3 Da, Purity >95%. ATP synthesis inhibitor. Antiprotozoal, antibiotic and antifungal agent. Shows antitumor effects against different cancer cell lines and neuroprotective effects.
The product is subject to the following: Abcam Restricted Use Statement