Learn More
Apexbio Technology LLC Bikinin(Synonyms: Bikinin, GSK3 Inhibitor VII, Glycogen Synthase Kinase-3 Inhibitor VII, GSK-3 Inhibitor VII), 10mM (in 1mL DMSO), CAS: 188011-69-0.
SDP
Catalog No. 5000060421
Encompass_Preferred
Bikinin (CAS 188011-69-0) is a small molecule inhibitor targeting glycogen synthase kinase 3 (GSK3)-like kinases particularly BIN2 thereby activating brassinosteroid (BR) signaling pathways downstream of the BRI1 receptor BR signaling regulates plant developmental processes including stem elongation vascular cell differentiation leaf expansion and stress responses Treatment with Bikinin can phenotypically rescue BR-insensitive mutants in Arabidopsis by restoring BR-dependent gene expression Additionally Bikinin usage promotes xylem cell differentiation in Nicotiana benthamiana leaf disk culture suggesting utility across different biological models
Catalog No. 50-000-60421 Supplier Apexbio Technology LLC Supplier No. B48985.1
Bikinin (CAS 188011-69-0) is a small molecule inhibitor targeting glycogen synthase kinase 3 (GSK3)-like kinases particularly BIN2 thereby activating brassinosteroid (BR) signaling pathways downstream of the BRI1 receptor BR signaling regulates plant developmental processes including stem elongation vascular cell differentiation leaf expansion and stress responses Treatment with Bikinin can phenotypically rescue BR-insensitive mutants in Arabidopsis by restoring BR-dependent gene expression Additionally Bikinin usage promotes xylem cell differentiation in Nicotiana benthamiana leaf disk culture suggesting utility across different biological models