Learn More
Medchemexpress LLC Bikinin (Abrasin) | 188011-69-0 | MFCD00116442 | 100.0% | 273.08 g/mol | C9H9BrN2O3 | 10MM 1ML
SDP
Catalog No. 5000235662
Encompass_Preferred
Bikinin is a non-steroidal, ATP-competitive inhibitor of plant GSK-3/Shaggy-like kinases that activates brassinosteroid (BR) signaling. It is supplied for research use in solution and solid forms, including a ready-to-use 10 mM solution in DMSO, and is reported at high purity for biochemical and plant signaling studies.
  • Non-steroidal ATP-competitive inhibitor of plant GSK-3/Shaggy-like kinases.
  • Activates brassinosteroid signaling pathways.
  • Available as a 10 mM solution in DMSO and as solid packs for custom preparation.
  • Reported high purity (99.95%) suitable for research applications.
  • Documentation available: datasheet, certificate of analysis, safety data sheet, and spectral data.
Catalog No. 50-002-35662 Supplier Medchemexpress LLC Supplier No. HY1252410MM 1ML
Bikinin is a non-steroidal, ATP-competitive inhibitor of plant GSK-3/Shaggy-like kinases that activates brassinosteroid (BR) signaling. It is supplied for research use in solution and solid forms, including a ready-to-use 10 mM solution in DMSO, and is reported at high purity for biochemical and plant signaling studies.
  • Non-steroidal ATP-competitive inhibitor of plant GSK-3/Shaggy-like kinases.
  • Activates brassinosteroid signaling pathways.
  • Available as a 10 mM solution in DMSO and as solid packs for custom preparation.
  • Reported high purity (99.95%) suitable for research applications.
  • Documentation available: datasheet, certificate of analysis, safety data sheet, and spectral data.