Learn More
Medchemexpress LLC Bilastine-d6 | 1215358-58-9 | 97.1% | C28H31D6N3O3 | 1 MG
SDP
Catalog No. 5000354484
Encompass_Preferred
Bilastine-d6 is a deuterium-labeled Bilastine. Bilastine is a selective histamine H1 receptor antagonist used for the treatment of allergic rhinoconjunctivitis and urticaria.
  • Used as a tracer.
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-54484 Supplier Medchemexpress LLC Supplier No. HY14447S1MG
Bilastine-d6 is a deuterium-labeled Bilastine. Bilastine is a selective histamine H1 receptor antagonist used for the treatment of allergic rhinoconjunctivitis and urticaria.
  • Used as a tracer.
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.