Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bilastine-d6 | 1215358-58-9 | MFCD34567868 | 97.1% | 469.65 | C28H31D6N3O3 | 5 MG
SDP

Catalog No. 5000221093
Encompass_Preferred

Bilastine-d6 is the deuterium-labeled analogue of bilastine, a selective histamine H1 receptor antagonist. Supplied for research use as a tracer or internal standard, it is intended for analytical and pharmacokinetic studies requiring isotopic labeling.

  • Deuterium-labeled analogue of bilastine.
  • Selective histamine H1 receptor antagonist profile.
  • Intended for research use and analytical applications.
  • Suitable as an internal standard for LC-MS and NMR quantitation.
  • Purity 97.12% (as supplied).
  • Soluble in DMSO at 10 mg/mL; requires ultrasonic and warming.
  • Offered in small research sizes (1 mg, 5 mg; larger sizes by quote).

Catalog No. 50-002-21093 Supplier Medchemexpress LLC Supplier No. HY14447S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bilastine-d6 is the deuterium-labeled analogue of bilastine, a selective histamine H1 receptor antagonist. Supplied for research use as a tracer or internal standard, it is intended for analytical and pharmacokinetic studies requiring isotopic labeling.

  • Deuterium-labeled analogue of bilastine.
  • Selective histamine H1 receptor antagonist profile.
  • Intended for research use and analytical applications.
  • Suitable as an internal standard for LC-MS and NMR quantitation.
  • Purity 97.12% (as supplied).
  • Soluble in DMSO at 10 mg/mL; requires ultrasonic and warming.
  • Offered in small research sizes (1 mg, 5 mg; larger sizes by quote).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.