Learn More
TARGETMOL CHEMICALS INC BILB-1941 5MG
SDP
Catalog No. 502580671
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BILB-1941 (BILB-1941ZW) is an inhibitor of HCV NS5B polymerase and can be used in studies about HCV infection. purity: 99%
Catalog No. 50-258-0671 Supplier TARGETMOL CHEMICALS INC Supplier No. T268155MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BILB-1941 (BILB-1941ZW) is an inhibitor of HCV NS5B polymerase and can be used in studies about HCV infection. purity: 99%