Learn More
Cayman Chemical BIle ACD Nuclear RECPTR FX 1ea
SDP
Catalog No. 502893420
Encompass_Preferred
Cayman’s Bile Acid Nuclear Receptor FXR (NR1H4) Reporter Assay Kit consists of a 96-well plate coated with a transfection complex containing DNA constructs for expressing FXR and a BSEP promoter-regulated SEAP reporter (FXR Reverse Transfection Strip Plate).
Catalog No. 50-289-3420 Supplier Cayman Chemical Supplier No. 6017901 EA
Cayman’s Bile Acid Nuclear Receptor FXR (NR1H4) Reporter Assay Kit consists of a 96-well plate coated with a transfection complex containing DNA constructs for expressing FXR and a BSEP promoter-regulated SEAP reporter (FXR Reverse Transfection Strip Plate).