Learn More
Sigma Aldrich Fine Chemicals Biosciences Bile Acid Assay Kit suffic
Catalog No. 502904062
Encompass
Twelve different types of bile acids are typically found in mammals among them two primary types are cholic acid and chenodeoxycholic acid. These can be dehydroxylated into secondary bile acids. finally these four can be conjugated to either taurine or glycine creating 8 different conjugated bile acids. Bile acid levels in feces blood urine and bile can be used as markers for various diseases such as hyperlipidemia cholestasis gall stones colon cancer etc. Bile acids also exist as sulfate salt forms known as bile acid sulfates. Sulfation of bile acids increases their solubility and decreases intestinal absorption thereby enhancing fecal and urinary excretion.
Catalog No. 50-290-4062 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. MAK3091KT
Twelve different types of bile acids are typically found in mammals among them two primary types are cholic acid and chenodeoxycholic acid. These can be dehydroxylated into secondary bile acids. finally these four can be conjugated to either taurine or glycine creating 8 different conjugated bile acids. Bile acid levels in feces blood urine and bile can be used as markers for various diseases such as hyperlipidemia cholestasis gall stones colon cancer etc. Bile acids also exist as sulfate salt forms known as bile acid sulfates. Sulfation of bile acids increases their solubility and decreases intestinal absorption thereby enhancing fecal and urinary excretion.