Learn More
Sigma Aldrich Fine Chemicals Biosciences Bile Acid/Carnitine/Sterol1EA
Catalog No. 502907055
Encompass
Compounds can either be used as standards and injected individually or a strategy may be employed to pool compounds to allow multiple mixtures to be analyzed per injection.
Catalog No. 50-290-7055 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. BACSMLS1EA
Compounds can either be used as standards and injected individually or a strategy may be employed to pool compounds to allow multiple mixtures to be analyzed per injection.