Learn More
Oxyrase Oxypras Bile Esculin Agar 10/Pk
SDP
Catalog No. NC9696625
Encompass
Innovative and specifically made for anaerobes to improve recovery of injured cells. Arrive reduced and ready to use; stays reduced in air for hours. Saves tech time; helps eliminate errors. Store refrigerated for a extended shelf life. Meets Best Practice criteria for working with clinical anaerobes. This product consists of 10 individually wrapped plates
Catalog No. NC9696625 Supplier Oxyrase Supplier No. PBBENB
Innovative and specifically made for anaerobes to improve recovery of injured cells. Arrive reduced and ready to use; stays reduced in air for hours. Saves tech time; helps eliminate errors. Store refrigerated for a extended shelf life. Meets Best Practice criteria for working with clinical anaerobes. This product consists of 10 individually wrapped plates