Learn More
MOLTOX Molecular Toxicology Inc Bile Esculin Azide Agar, 20 mL/plate, 10 plates/sleeve, Store in Refrigerator
SDP
Catalog No. 50948806
Encompass_Preferred
Also known as Enterococcosel Agar, this medium is used as a rapid selective medium for the detection and enumeration of enterococci. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-806 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 5140S006
Also known as Enterococcosel Agar, this medium is used as a rapid selective medium for the detection and enumeration of enterococci. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.