Learn More
NEOGEN Bile Esculin Azide Agar, 10 kg
SDP
Catalog No. 502015635
Encompass_Preferred
Bile Esculin Azide Agar is used for the selective isolation and differentiation of group D streptococci in a laboratory setting. Bile Esculin Azide Agar is not intended for use in the diagnosis of disease or other conditions in humans.Bile Esculin Azide Agar is a modification of the medium reported by Isenberg and Isenberg, Goldberg, and Sampson. This formula modifies Bile Esculin Agar by adding sodium azide and reducing the concentration of bile. The revised medium is more selective, but still provides rapid growth and efficient recovery of group D streptococci.Molecular taxonomic studies of the genus Streptococcus have placed enterococci, previously described group D streptococci, in the genus Enterococcus. The ability to hydrolyze esculin in the presence of bile is a characteristic of enterococci and group D streptococci.For more information visit https://www.neogen.com/solutions/Microbiology/bile-esculin-azide-agar
Catalog No. 50-201-5635 Supplier NEOGEN Supplier No. 700003458
Bile Esculin Azide Agar is used for the selective isolation and differentiation of group D streptococci in a laboratory setting. Bile Esculin Azide Agar is not intended for use in the diagnosis of disease or other conditions in humans.Bile Esculin Azide Agar is a modification of the medium reported by Isenberg and Isenberg, Goldberg, and Sampson. This formula modifies Bile Esculin Agar by adding sodium azide and reducing the concentration of bile. The revised medium is more selective, but still provides rapid growth and efficient recovery of group D streptococci.Molecular taxonomic studies of the genus Streptococcus have placed enterococci, previously described group D streptococci, in the genus Enterococcus. The ability to hydrolyze esculin in the presence of bile is a characteristic of enterococci and group D streptococci.For more information visit https://www.neogen.com/solutions/Microbiology/bile-esculin-azide-agar