Learn More
Instech Loop connector for dual channel rat Vascular Access Harness. Sterile. Shunts bile from one channel back to the other. Use with VAHD115AB harness.
SDP
Catalog No. 5081256
Encompass
The Instech VAHD115L is a sterile loop connector designed for use with dual channel VAH harnesses in rats, creating a closed shunt that routes bile from one channel back to the other during non‑sampling periods. When paired with the VAHD115AB harness, it maintains natural bile flow and catheter patency until the loop is removed and a tether is attached for collection or infusion procedures.
Features
  • Sterile loop connector for dual channel rat VAH systems
  • Shunts bile from one channel to the other when not sampling
  • Designed for use with VAHD115AB Vascular Access Harness™
  • Maintains catheter patency in bile sampling workflows
  • Quick connection and removal for study transitions
Catalog No. 50-812-56 Supplier Instech Supplier No. VAHD115L
The Instech VAHD115L is a sterile loop connector designed for use with dual channel VAH harnesses in rats, creating a closed shunt that routes bile from one channel back to the other during non‑sampling periods. When paired with the VAHD115AB harness, it maintains natural bile flow and catheter patency until the loop is removed and a tether is attached for collection or infusion procedures.
Features
  • Sterile loop connector for dual channel rat VAH systems
  • Shunts bile from one channel to the other when not sampling
  • Designed for use with VAHD115AB Vascular Access Harness™
  • Maintains catheter patency in bile sampling workflows
  • Quick connection and removal for study transitions