Learn More
Supelco Inc Bile salts, 50 G
Catalog No. NC0063082
Encompass
For microbiology a mixture of sodium cholate and sodium deoxycholate. Bile salts may be used as one of the selective agents to prevent the growth of injured Escherichia coli cells, in an experiment done in order to identify the specific agent in the selective media which would inhibit the growth of the injured Escherichia coli ML30 cells. It may also be used for identification of isolates of Vibrio vulnificus which is a halophilic bacterium, found responsible for causing several human infections.
Catalog No. NC0063082 Supplier Supelco Inc Supplier No. B875650G
For microbiology a mixture of sodium cholate and sodium deoxycholate. Bile salts may be used as one of the selective agents to prevent the growth of injured Escherichia coli cells, in an experiment done in order to identify the specific agent in the selective media which would inhibit the growth of the injured Escherichia coli ML30 cells. It may also be used for identification of isolates of Vibrio vulnificus which is a halophilic bacterium, found responsible for causing several human infections.