Learn More
Research Products International Corp Bile Salts No. 3 1 KG
SDP
Catalog No. 502230364
Encompass_Preferred
Bile Salts No. 3 is refined bile salt, comprising mainly of sodium cholate and sodium deoxycholate. It is used in media as a agent to isolate enteric organisms and coliforms. It is an excellent inhibitor of gram positive bacteria such as streptococci and staphylocci.
Catalog No. 50-223-0364 Supplier Research Products International Corp Supplier No. B117501000.0
Bile Salts No. 3 is refined bile salt, comprising mainly of sodium cholate and sodium deoxycholate. It is used in media as a agent to isolate enteric organisms and coliforms. It is an excellent inhibitor of gram positive bacteria such as streptococci and staphylocci.