Learn More
Research Products International Corp Bile Salts No. 3 10 KG
SDP
Catalog No. 502230363
Encompass_Preferred
Bile Salts No. 3 is refined bile salt, comprising mainly of sodium cholate and sodium deoxycholate. It is used in media as a agent to isolate enteric organisms and coliforms. It is an excellent inhibitor of gram positive bacteria such as streptococci and staphylocci.
Catalog No. 50-223-0363 Supplier Research Products International Corp Supplier No. B1175010000.0
Add to Cart
Add to Cart
Request a Quote
Bile Salts No. 3 is refined bile salt, comprising mainly of sodium cholate and sodium deoxycholate. It is used in media as a agent to isolate enteric organisms and coliforms. It is an excellent inhibitor of gram positive bacteria such as streptococci and staphylocci.