Learn More
Research Products International Corp Bile Salts Number 3, 25 Grams
SDP
Catalog No. 50995794
Encompass_Preferred
Bile Salts No. 3 is refined bile salt, comprising mainly of sodium cholate and sodium deoxycholate. It is used in media as a agent to isolate enteric organisms and coliforms. It is an excellent inhibitor of gram positive bacteria such as streptococci and staphylocci.
Catalog No. 50-995-794 Supplier Research Products International Corp Supplier No. B1175025.0
Bile Salts No. 3 is refined bile salt, comprising mainly of sodium cholate and sodium deoxycholate. It is used in media as a agent to isolate enteric organisms and coliforms. It is an excellent inhibitor of gram positive bacteria such as streptococci and staphylocci.