Learn More
Alkali Scientific Bile Salts No. 3, 5 Kg
SDP
Catalog No. 5000659683
Encompass_Preferred
Bile Salts No. 3 is a high-quality reagent primarily used in laboratory applications for culturing and isolating microorganisms. Available in a 5 Kg package, this standard-grade reagent is ideal for use in the preparation of selective media, particularly for microbial growth and differentiation. Bile Salts No. 3 helps promote the growth of bile-tolerant organisms while inhibiting the growth of others, making it a key ingredient in selective media for enteric bacteria. This product is commonly used in microbiological, clinical, and research laboratories where accurate and consistent results are essential for microbial culture and testing protocols.
Catalog No. 50-006-59683 Supplier Alkali Scientific Supplier No. ILE002CP
Bile Salts No. 3 is a high-quality reagent primarily used in laboratory applications for culturing and isolating microorganisms. Available in a 5 Kg package, this standard-grade reagent is ideal for use in the preparation of selective media, particularly for microbial growth and differentiation. Bile Salts No. 3 helps promote the growth of bile-tolerant organisms while inhibiting the growth of others, making it a key ingredient in selective media for enteric bacteria. This product is commonly used in microbiological, clinical, and research laboratories where accurate and consistent results are essential for microbial culture and testing protocols.