Learn More
Alkali Scientific Bile Salts No. 3, Microbiology Grade, 10 Kg
SDP
Catalog No. 5000659654
Encompass_Preferred
Bile Salts No. 3, Microbiology Grade, 10kg is a bulk reagent used for preparing microbiological media designed to isolate and differentiate enteric bacteria. The high purity of these bile salts makes them essential for selective bacterial growth in food safety, clinical microbiology, and environmental monitoring. This large quantity is ideal for laboratories that require frequent preparation of selective media in large volumes. Bile Salts No. 3 works by inhibiting the growth of non-enteric bacteria while allowing enteric pathogens such as Salmonella, Shigella, and Escherichia coli to thrive. It is commonly used in bacteriological testing of water, food, and clinical specimens.
Catalog No. 50-006-59654 Supplier Alkali Scientific Supplier No. ILE001CP
Bile Salts No. 3, Microbiology Grade, 10kg is a bulk reagent used for preparing microbiological media designed to isolate and differentiate enteric bacteria. The high purity of these bile salts makes them essential for selective bacterial growth in food safety, clinical microbiology, and environmental monitoring. This large quantity is ideal for laboratories that require frequent preparation of selective media in large volumes. Bile Salts No. 3 works by inhibiting the growth of non-enteric bacteria while allowing enteric pathogens such as Salmonella, Shigella, and Escherichia coli to thrive. It is commonly used in bacteriological testing of water, food, and clinical specimens.