Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Bilingual Chemical, Biohazard & Hazardous Material Sign: DANGER KEEP ALL CYLINDERS CHAINED

Catalog No. 19105107 Shop All Brady Corporation Products
Change view
Click to view available options
Dimensions (L x W):
14 x 10 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19-105-107 14 x 10 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-105-107 Supplier Brady™ Supplier No. 125258

missing translation for 'validMainframeMsg'

Chemical, Biohazard & Hazardous Material Sign - Self-Sticking Polyester, English and Spanish Bilingual: Self-Sticking Polyester (B-302) is a flexible polymer plastic over-laminated with a clear plastic coating to protect graphics against abrasion, sunlight, chemicals.

  • Weathers up to 8 years outdoors, even longer indoors, and withstands wide temperature variations (up to 230°F)
  • Flexible material conforms to most surfaces while protected graphics hold up to harsh environments, abrasion, and repeated spills and cleanings
  • Permanent, pressure sensitive adhesive forms a long-lasting bond to even the most difficult surfaces and shapes
  • Cold temperature adhesive allows the signage to be applied at 0°F (-18°C)

Specifications

Certifications/Compliance Globally Harmonized System of Classification and Labeling of Chemicals (GHS); NFPA 704; OSHA 1910.1200
Finish Gloss
Material Polyester
Quantity 1 Ea.
Adhesive Type Permanent Acrylic
Resistant To Abrasion-Resistant; Chemical-Resistant
Mounting Type Self-adhesive
For Use With (Application) Hazard Material Safety
Height (English) 14 in.
Height (Metric) 35.56 cm
Thickness 0.01 in.
Width (English) 10 in.
Width (Metric) 25.4 cm
Temperature Range (Metric) -40 °C to 110 °C
Shape Rectangular
Temperature Range (English) -40 °F to 230 °F
Languages English, Spanish
Dimensions (L x W) 14 x 10 in.
Displayed Text KEEP ALL CYLINDERS CHAINED/MANTENER TODOS LOS CILINDROS ENCADENADOS
Product Type Sign
Color Black
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.