Learn More
Description
Helps prevent costly mistakes by providing potentially life-saving information to employees
Specifications
Specifications
| Format | Wall Mounted |
| Product Type | Information Center |
| Width (Metric) | 660.4 mm |
| Quantity | Each |
| Languages | English, Spanish |
| For Use With (Application) | Hazard Communication |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.