Learn More
Marketlab Bilingual STAT Bags
Catalog No. 22505011
Change view
Click to view available options
Quantity:
100/Pk.
Catalog No. Quantity
22-505-011 100/Pk.
1 options
Catalog No. 22-505-011 Supplier Marketlab Supplier No. 108164
Add to Cart
Add to Cart
Bilingual labeling helps improve STAT specimen identification. Designed for clear communication and handling.
  • Printed with key information in English and Spanish
  • All-red bag with bold, black lettering and biohazard symbol for easy identification
  • Reclosable zipper seal for fast, easy, and liquid-tight closure
  • Easy-open tear line improves turn-around time
  • Three-wall construction keeps paperwork and specimen separate
  • 6 W x 9 H in.
Specifications
Autoclavable Not Autoclavable
Closure Type Zip Closure
Color Red with Black Print
Disposable Single-use
Material LDPE
Sterility Non-sterile
Quantity 100/Pk.
Length (English) 9 in.
Width (English) 6 in.
Product Type Specimen Bag