Learn More
Sigma Aldrich Fine Chemicals Biosciences Bilirubin Assay Kit suffic
Catalog No. 502904890
Encompass
Bilirubin also known as hematoidin is a degradation product formed as a result of heme catabolism in the liver. Bilirubin circulates in the blood stream as either the unconjugated insoluble form (indirect bilirubin) or the soluble glucuronide-conjugated form (direct bilirubin). Conjugated bilirubin moves from the bile canaliculi of the liver to the gall bladder where it is excreted into the small intestine during digestion. High levels of bilirubin can result in jaundice and may indicate liver disease blood disorders or blockage of the bile ducts.
Catalog No. 50-290-4890 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. MAK1261KT
Bilirubin also known as hematoidin is a degradation product formed as a result of heme catabolism in the liver. Bilirubin circulates in the blood stream as either the unconjugated insoluble form (indirect bilirubin) or the soluble glucuronide-conjugated form (direct bilirubin). Conjugated bilirubin moves from the bile canaliculi of the liver to the gall bladder where it is excreted into the small intestine during digestion. High levels of bilirubin can result in jaundice and may indicate liver disease blood disorders or blockage of the bile ducts.